Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC1A1, Mouse anti-Human, ATTO565, Clone: S180-41, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB18289

Catalog No. 89-338-214


Add to cart

Description

Description

Specifications

Specifications

SLC1A1
S180-41
Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user.
solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1
EAAC1|EAAT3
Mouse
IgG1
RUO
Antibody
Monoclonal
Human, Rat
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
ATTO565
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Liquid
SLC1A1
Recombinant protein corresponding to full length human SLC1A1.
100 μg
Store at -20°C.
Primary
6505
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit