Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC20A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542142
Description
Peptide sequence: DVNLYNETVE TLMAGEVSAM VGSAVWQLIA SFLRLPISGT HCIVGSTIGF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
This gene encodes a member of the inorganic phosphate transporter family. The encoded protein is a type 3 sodium-dependent phosphate symporter that plays an important role in phosphate homeostasis by mediating cellular phosphate uptake. The encoded protein also confers susceptibility to viral infection as a gamma-retroviral receptor. Mutations in this gene may play a role in familial idiopathic basal ganglia calcification. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
SLC20A2 | |
Polyclonal | |
Unconjugated | |
SLC20A2 | |
Ab1-188; fi23g11; Gibbon ape leukemia virus receptor 2; Glvr2; GLVR-2; hPit2; IBGC1; IBGC3; MLVAR; MolPit2; murine leukemia virus, amphotropic, receptor for; murine leukemia virus, amphotropic; receptor; phosphate transporter 2; PIT2; PIT-2; RAM1; RAM-1; receptor for amphitropic viruses 1; receptor for amphotropic viruses 1; slc20a2; Sodium-dependent phosphate transporter 2; solute carrier family 20 (phosphate transporter), member 2; solute carrier family 20 member 2; solute carrier family 20, member 2; type III sodium-dependent phosphate transporter; wu:fi23g11; zgc:152990 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6575 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q08357 | |
SLC20A2 | |
Synthetic peptide directed towards the N-terminal of human SLC20A2 (aa 76-125). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction