Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC22A15 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen SLC22A15
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095218
View Documents
Novus Biologicals
NBP305043100UL
100 μg
Each for $320.71
Add to cart
 
NB095219
View Documents
Novus Biologicals
NBP30504320UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

SLC22A15 Polyclonal antibody specifically detects SLC22A15 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

SLC22A15
Unconjugated
Rabbit
Human, Mouse
DKFZp761G0313, Flipt 1, FLIPT1PRO34686, fly-like putative organic ion transporter 1, Fly-like putative transporter 1, solute carrier family 22 (organic cation transporter), member 15, solute carrier family 22 member 15, solute carrier family 22, member 15, trans-like protein
Recombinant fusion protein containing a sequence corresponding to amino acids 443-547 of human SLC22A15 (NP_060890.2).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
55356
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit