Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC22A17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598649
Description
SLC22A17 Polyclonal Antibody for ICC/IF, IHC (P), ELISA
The Slc22 family of organic anion and cation transporters (OATs, OCTs, OCTNs) are transmembrane proteins expressed predominantly in kidney and liver. Each contain 12 predicted alpha-helical transmembrane domains (TMDs) and one large extracellular loop between TMDs 1 and 2. Transporters of the SLC22 family function in different ways such as uniporters that mediate facilitated diffusion in either direction (OCTs), as anion exchangers (OAT1, OAT3 and URAT1), and as Na(+)/l-carnitine cotransporter (OCTN2). Slc22 family members participate in the absorption and/or excretion of drugs, xenobiotics, and endogenous compounds in intestine, liver, and kidney, and perform homeostatic functions in brain and heart. Mutations in the Slc22 family may cause specific diseases such as primary systemic carnitine deficiency or idiopathic renal hypouricemia and may change drug absorption or excretion. Recent studies show the expression of Slc22A17 as receptor for Lipocalin 2 is relatively high in hematopoietic stem cells.
Specifications
SLC22A17 | |
Polyclonal | |
Unconjugated | |
SLC22A17 | |
1700094C23Rik; 24p3 receptor; 24p3R; AU041908; AW555662; BOCT; Boit; brain-specific organic ion transporter; brain-type organic cation transporter; hBOIT; Lipocalin-2 receptor; mBOCT; Neutrophil gelatinase-associated lipocalin receptor; NGAL receptor; NgalR; NGALR2; NGALR3; organic cation transporter BOCT; potent brain type organic ion transporter; rBOCT; SLC22A17; solute carrier family 22 (organic cation transporter), member 17; solute carrier family 22 member 17; solute carrier family 22, member 17 | |
Rabbit | |
Protein G | |
RUO | |
51310 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8WUG5 | |
SLC22A17 | |
Recombinant Human Solute carrier family 22 member 17 protein (1-99AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction