Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC22A18AS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531339
Description
Recommended positive controls: 293T, A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The SLC22A18/SLC22A18AS genes are a sense-antisense pair located at human chromosome segment 11p15.5. These genes are paternally imprinted: paternal alleles are silenced and maternal alleles are expressed. SLC22A18AS (antisense) is associated to SLC22A18 (sense). These two genes share, in divergent orientations, 31 bp in their 5' regions (between the first exon of SLC22A18AS and the second exon of SLC22A18).
Specifications
SLC22A18AS | |
Polyclonal | |
Unconjugated | |
SLC22A18AS | |
Beckwith-Wiedemann region 1B; beckwith-Wiedemann syndrome chromosomal region 1 candidate gene B protein; Beckwith-Wiedemann syndrome chromosome region 1, candidate b; BWR1B; BWSCR1B; ORCTL2S; organic cation transporter-like 2 antisense; organic cation transporter-like protein 2 antisense protein; p27-Beckwith-Wiedemann region 1 B; p27-BWR1B; SLC22A18AS; SLC22A1LS; solute carrier family 22 (organic cation transporter), member 18 antisense; solute carrier family 22 (organic cation transporter), member 1-like antisense; solute carrier family 22 member 18 antisense; solute carrier family 22 member 18 antisense protein; solute carrier family 22 member 1-like antisense protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5003 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.75 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8N1D0 | |
SLC22A18AS | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 139 of Human SLC22A18AS. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction