Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC23A3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen SLC23A3
Immunogen Synthetic peptides corresponding to SLC23A3(solute carrier family 23 (nucleobase transporters), member 3) The peptide sequence was selected from the N terminal of SLC23A3. Peptide sequence SRSPLNPSQLRSVGSQDALAPLPPPAPQNPSTHSWDPLCGSLPWGLSCLL
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15989620 View Documents Novus BiologicalsSupplier Diversity Partner
NBP15989620UL
20ul Each for $149.00
Add to cart
 
NBP159896 View Documents Novus BiologicalsSupplier Diversity Partner
NBP159896
100 ul Each for $329.00
Add to cart
 
Description

Description

SLC23A3 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

SLC23A3
Rabbit
Polyclonal
Unconjugated
151295
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to SLC23A3(solute carrier family 23 (nucleobase transporters), member 3) The peptide sequence was selected from the N terminal of SLC23A3. Peptide sequence SRSPLNPSQLRSVGSQDALAPLPPPAPQNPSTHSWDPLCGSLPWGLSCLL
Primary
IgG
E2-binding protein 3, E2BP3, FLJ31168, Na(+)/L-ascorbic acid transporter 3, Sodium-dependent vitamin C transporter 3, solute carrier family 23 (nucleobase transporters), member 3, solute carrier family 23 member 3, SVCT3HPC E2-binding protein 3, Yspl1
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit