Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC24A1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen SLC24A1
Immunogen Synthetic peptides corresponding to SLC24A1(solute carrier family 24 (sodium/potassium/calcium exchanger), member 1) The peptide sequence was selected from the middle region of SLC24A1. Peptide sequence EQLSRRPVAKVMALEDLSKPGDGAIAVDELQDNKKL
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1625220 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16252120UL
20ul Each for $149.00
Add to cart
 
NBP162521 View Documents Novus BiologicalsSupplier Diversity Partner
NBP162521
100 ul Each for $329.00
Add to cart
 
Description

Description

SLC24A1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

SLC24A1
Rabbit
Polyclonal
Unconjugated
9187
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to SLC24A1(solute carrier family 24 (sodium/potassium/calcium exchanger), member 1) The peptide sequence was selected from the middle region of SLC24A1. Peptide sequence EQLSRRPVAKVMALEDLSKPGDGAIAVDELQDNKKL
Primary
IgG
HsT17412, KIAA0702Na(+)/K(+)/Ca(2+)-exchange protein 1, NCKX, NCKX1Retinal rod Na-Ca+K exchanger, retinal rod Na+/Ca+/K+ exchanger, RODX, sodium/potassium/calcium exchanger 1, solute carrier family 24 (sodium/potassium/calcium exchanger), member 1
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit