Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC24A5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen SLC24A5
Applications Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096149
View Documents
Novus Biologicals
NBP304861100UL
100 μg Each for $320.71
Add to cart
 
NB096150
View Documents
Novus Biologicals
NBP30486120UL
20 μg Each for $138.37
Add to cart
 
Description

Description

SLC24A5 Polyclonal antibody specifically detects SLC24A5 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

SLC24A5
Unconjugated
Rabbit
Human
ion transporter JSX, JSXsodium/potassium/calcium exchanger 5, Na(+)/K(+)/Ca(2+)-exchange protein 5, NCKX5solute carrier family 24 (sodium/potassium/calcium exchanger), member 5, SHEP4, Solute carrier family 24 member 5, solute carrier family 24, member 5
Recombinant fusion protein containing a sequence corresponding to amino acids 220-300 of human SLC24A5 (NP_995322.1).
Polyclonal
Affinity purified
Immunofluorescence
Purified
RUO
PBS with 50% glycerol, pH7.3.
283652
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit