Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC25A11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542428
Description
Peptide sequence: AATASAGAGG IDGKPRTSPK SVKFLFGGLA GMGATVFVQP LDLVKNRMQL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 85%.
The oxoglutarate/malate carrier transports 2-oxoglutarate across the inner membranes of mitochondria in an electroneutral exchange for malate or other dicarboxylic acids (summary by Iacobazzi et al., 1992 [PubMed 1457818]).
Specifications
SLC25A11 | |
Polyclonal | |
Unconjugated | |
SLC25A11 | |
2310022P18Rik; 2oxoc; 2-oxoglutarate carrier; 2-oxoglutarate/malate carrier; 2-oxoglutarate/malate carrier protein; fa07h09; Mitochondrial 2-oxoglutarate/malate carrier protein; OGC; OGCP; OMC; SLC20A4; SLC25A11; solute carrier family 20 (oxoglutarate carrier), member 4; solute carrier family 25 (mitochondrial carrier oxoglutarate carrier), member 11; solute carrier family 25 (mitochondrial carrier; oxoglutarate carrier), member 11; solute carrier family 25 member 11; wu:fa07h09; zgc:86898 | |
Rabbit | |
Affinity chromatography | |
RUO | |
8402 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q02978 | |
SLC25A11 | |
Synthetic peptide directed towards the N-terminal of human SLC25A11 (aa 2-51). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction