Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC25A17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116600
Description
Positive Samples: Mouse liver, Mouse kidney Immunogen sequence: IDAFHQIIRD EGISALWNGT FPSLLLVFNP AIQFMFYEGL KRQLLKKRMK LSSLDVFIIG AVAKAIATTV TYPLQTVQSI LRFGRHRLNP ENRTLGSLRN I.
This protein encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Studies suggest that this protein may regulate cytokine production, and thus play a key role in the control of the inflammatory response.
Specifications
SLC25A17 | |
Polyclonal | |
Unconjugated | |
SLC25A17 | |
34 kDa peroxisomal membrane protein; 34kDa; 47kDa; peroxisomal integral membrane protein 47; peroxisomal membrane protein PMP34; peroxisomal membrane protein, 34 kDa; Pmp34; Pmp35; Pmp47; SLC25A17; solute carrier family 25 (mitochondrial carrier, peroxisomal membrane protein), member 17; solute carrier family 25 (mitochondrial carrier; peroxisomal membrane protein, 34kDa), member 17; solute carrier family 25 member 17; solute carrier family 25, member 17 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10478, 20524 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.92 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O43808, O70579 | |
SLC25A17 | |
A synthetic peptide corresponding to a sequence within amino acids 150-250 of human SLC25A17 (NP_0063491). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction