Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A18 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | SLC25A18 |
---|---|
Dilution | Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SLC25A18 Polyclonal antibody specifically detects SLC25A18 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
SLC25A18 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
GC-2, GC2Glutamate/H(+) symporter 2, mitochondrial glutamate carrier 2, solute carrier family 25 (mitochondrial carrier), member 18, Solute carrier family 25 member 18 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: NDFFRRLLMEDGMQRNLKMEM | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
83733 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title