Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550790

Catalog No. PA550790


Add to cart

Description

Description

The antibody detects endogenous levels of total SLC25A2 protein.

Located between the protocadherin beta and gamma gene clusters on chromosome 5, this intronless gene encodes a protein that is highly similar to an ornithine transporter localized in the mitochondrial inner membrane. The encoded protein most likely plays a role in metabolism as a mitochondrial transport protein.
Specifications

Specifications

SLC25A2
Polyclonal
Unconjugated
SLC25A2
Liquid
SLC25A2
Fusion protein corresponding to residues near the N terminal of human solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.2 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9BXI2
ORNT2
Rabbit
IgG
100 μL
-20°C
Primary
83884
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit