Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309473100UL
Description
SLC25A2 Polyclonal specifically detects SLC25A2 in Human samples. It is validated for Western Blot.Specifications
SLC25A2 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
mitochondrial ornithine transporter 2, ORC2, ORNT2, solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 2 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human SLC25A2 (NP_114153). Peptide sequence VPGYFFFFGGYELSRSFFASGRSKDELGPVHLMLSGGVAGICLWLVVFPV | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
83884 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction