Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen SLC25A20
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093354
View Documents
Novus Biologicals
NBP304656100UL
100 μg Each for $320.71
Add to cart
 
NB093355
View Documents
Novus Biologicals
NBP30465620UL
20 μg Each for $138.37
Add to cart
 
Description

Description

SLC25A20 Polyclonal antibody specifically detects SLC25A20 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

SLC25A20
Unconjugated
Rabbit
Human, Mouse, Rat
CACCarnitine/acylcarnitine translocase, CACTSolute carrier family 25 member 20, mitochondrial carnitine/acylcarnitine carrier protein, solute carrier family 25 (carnitine/acylcarnitine translocase), member 20
Recombinant fusion protein containing a sequence corresponding to amino acids 1-301 of human SLC25A20 (NP_000378.1).
Polyclonal
Affinity purified
Immunofluorescence, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
788
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit