Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | SLC25A22 |
---|---|
Applications | Western Blot, Immunofluorescence, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB022698
|
Novus Biologicals
NBP294069100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB022699
|
Novus Biologicals
NBP29406920UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
SLC25A22 Polyclonal antibody specifically detects SLC25A22 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, KnockDownSpecifications
SLC25A22 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.3), 50% glycerol | |
79751 | |
IgG | |
Affinity purified |
Western Blot, Immunofluorescence, KnockDown | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
EIEE3mitochondrial glutamate carrier 1, FLJ13044, GC1GC-1, Glutamate/H(+) symporter 1, NET44, solute carrier family 25 (mitochondrial carrier: glutamate), member 22, Solute carrier family 25 member 22 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 128-188 of human SLC25A22 (NP_078974.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title