Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC25A23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113375
Description
SLC25A23 Polyclonal Antibody for Western Blot, ELISA
Calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane (PubMed:15123600). May act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria (PubMed:15123600). Acts as a regulator of mitochondrial calcium uptake via interaction with MCU and MICU1 (PubMed:24430870). [UniProt].
Specifications
SLC25A23 | |
Polyclonal | |
Unconjugated | |
SLC25A23 | |
2310067G05Rik; APC2; calcium-binding mitochondrial carrier protein SCaMC-3; MCSC2; mitochondrial ATP-Mg/Pi carrier protein 2; Mitochondrial Ca(2+)-dependent solute carrier protein 2; mitochondrial Ca2+-dependent solute carrier protein 2; phosphate carrier), member 23; SCAMC3; SCaMC-3; short calcium-binding mitochondrial carrier 3; Slc25a23; sma; Small calcium-binding mitochondrial carrier protein 3; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 23; solute carrier family 25 member 23 | |
Rabbit | |
Protein G | |
RUO | |
66972, 79085 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q6GQS1, Q9BV35 | |
SLC25A23 | |
Recombinant Human Calcium-binding mitochondrial carrier protein SCaMC-3 protein (1-188AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction