Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A23 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP258927

Catalog No. NBP258927


Add to cart

Description

Description

SLC25A23 Polyclonal antibody specifically detects SLC25A23 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

SLC25A23
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
APC2FLJ30339, calcium-binding mitochondrial carrier protein SCaMC-3, MCSC2, MGC2615, Mitochondrial ATP-Mg/Pi carrier protein 2, Mitochondrial Ca(2+)-dependent solute carrier protein 2, mitochondrial Ca2+-dependent solute carrier protein 2, SCaMC-3, SCAMC3, short calcium-binding mitochondrial carrier 3, Small calcium-binding mitochondrial carrier protein 3, solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 23, Solute carrier family 25 member 23
Rabbit
IgG
100 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunocytochemistry, Immunofluorescence
Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Affinity Purified
SLC25A23
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ELDSNKDGRVDVHELRQGLARLGGGNPDPGAQQGISSEGDADPDGGLDLEEFSRYLQEREQRLLLMFHSLDRNQDGHIDVSEIQQSFRALGISISLEQAEKILHSMDRDGTMTIDWQEWRDHFLLHSL
Affinity purified
RUO
Primary
79085
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only