Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A24 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP257669

Catalog No. NBP257669


Add to cart

Description

Description

SLC25A24 Polyclonal antibody specifically detects SLC25A24 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

SLC25A24
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
APC1calcium-binding mitochondrial carrier protein SCaMC-1, calcium-binding transporter, DKFZp586G0123, MCSC1, Mitochondrial ATP-Mg/Pi carrier protein 1, mitochondrial ATP-Mg/Pi transporter, Mitochondrial Ca(2+)-dependent solute carrier protein 1, SCAMC1, SCAMC-1, short calcium-binding mitochondrial carrier 1, Small calcium-binding mitochondrial carrier protein 1, solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 24, Solute carrier family 25 member 24
Rabbit
IgG
100 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500
Affinity Purified
SLC25A24
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MLRWLRDFVLPTAACQDAEQPTRYETLFQALDRNGDGVVDIGELQEGLRNLGIPLGQDAEEKIFTTGDVNKDGKL
Affinity purified
RUO
Primary
29957
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only