Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A26 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550789
Description
The antibody detects endogenous levels of total SLC25A26 protein.
SLC25A26 Polyclonal antibody specifically detects SLC25A26 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
SLC25A26 | |
Polyclonal | |
Unconjugated | |
SLC25A26 | |
4930433D19Rik; 4933433F13Rik; AW557176; COXPD28; D6Bwg0781e; hypothetical protein LOC560478; mitochondrial S-adenosylmethionine transporter; S-adenosylmethionine mitochondrial carrier protein; samc; SLC25A26; Slc25a6; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 26; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 26; solute carrier family 25 (S-adenosylmethionine carrier), member 26; solute carrier family 25 member 26; solute carrier family 25, member 26; SZFULLNAME2; SZGENESYMBOL2; zgc:110080 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
115286 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q70HW3 | |
SLC25A26 | |
Fusion protein corresponding to residues near the C terminal of human solute carrier family 25 (S-adenosylmethionine carrier), member 26 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction