Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550761

Catalog No. PA550761


Add to cart

Description

Description

The antibody detects endogenous levels of total SLC25A3 protein.

The protein encoded by this gene catalyzes the transport of phosphate into the mitochondrial matrix, either by proton cotransport or in exchange for hydroxyl ions. The protein contains three related segments arranged in tandem which are related to those found in other characterized members of the mitochondrial carrier family. Both the N-terminal and C-terminal regions of this protein protrude toward the cytosol.
Specifications

Specifications

SLC25A3
Polyclonal
Unconjugated
SLC25A3
Liquid
SLC25A3
Synthetic peptide corresponding to residues near the C terminal of human solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
2.4 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q00325
PHC, PTP, OK/SW-cl.48
Rabbit
IgG
100 μL
-20°C
Primary
5250
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit