Learn More
SLC25A3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 158551AP
Catalog No. 50-173-0532
Add to cart
Description
Description
The protein encoded by this gene catalyzes the transport of phosphate into the mitochondrial matrix, either by proton cotransport or in exchange for hydroxyl ions. The protein contains three related segments arranged in tandem which are related to those found in other characterized members of the mitochondrial carrier family. Both the N-terminal and C-terminal regions of this protein protrude toward the cytosol.
Specifications
Specifications
SLC25A3
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q00325, Q8VEM8
OK/SW cl.48, PHC, phosphate carrier, Phosphate transport protein, PiC, PTP, SLC25A3
Rabbit
IgG
150 μL
-20°C
Primary
18674, 5250
Immunoprecipitation, Western Blot
Unconjugated
SLC25A3
Liquid
SLC25A3
SLC25A3 Fusion Protein Ag8698
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse