Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A31 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen SLC25A31
Immunogen Synthetic peptides corresponding to SLC25A31(solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 31) The peptide sequence was selected from the N terminal of SLC25A31. Peptide sequence LAGGVAAAVSKTAVAP
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1595620 View Documents Novus BiologicalsSupplier Diversity Partner
NBP15956120UL
20ul Each for $149.00
Add to cart
 
NBP159561 View Documents Novus BiologicalsSupplier Diversity Partner
NBP159561
100 ul Each for $329.00
Add to cart
 
Description

Description

SLC25A31 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

SLC25A31
Rabbit
Polyclonal
Unconjugated
83447
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to SLC25A31(solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 31) The peptide sequence was selected from the N terminal of SLC25A31. Peptide sequence LAGGVAAAVSKTAVAP
Primary
IgG
AAC4DKFZp434N1235, adenine nucleotide translocase 4, Adenine nucleotide translocator 4, ADP, ADP/ATP translocase 4, ANT 4, ANT4DKFZP434N1235, ATP carrier protein 4, SFEC, SFEC35kDa, solute carrier family 25 (mitochondrial carrier; adenine nucleotidetranslocator), member 31, Solute carrier family 25 member 31, Sperm flagellar energy carrier protein
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit