Learn More
SLC25A33 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 177941AP
Catalog No. 50-173-1795
Add to cart
Specifications
Specifications
SLC25A33
0.14 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BSK2
BMSC MCP, HuBMSC MCP, Protein PNC1, SLC25A33
Rabbit
IgG
150 μL
-20°C
Primary
84275
Western Blot
Unconjugated
SLC25A33
Liquid
SLC25A33
SLC25A33 Fusion Protein Ag11926
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human