Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A42 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15960520UL
Description
SLC25A42 Polyclonal specifically detects SLC25A42 in Human samples. It is validated for Western Blot.Specifications
SLC25A42 | |
Polyclonal | |
Western Blot 0.2-1 ug/ml | |
Q86VD7 | |
SLC25A42 | |
Synthetic peptides corresponding to SLC25A42(solute carrier family 25, member 42) The peptide sequence was selected from the N terminal of SLC25A42. Peptide sequence GNGVKEGPVRLHEDAEAVLSSSVSSKRDHRQVLSSLLPGALAGALAKTAV. | |
Affinity Purified | |
RUO | |
284439 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
MGC26694, solute carrier family 25 member 42, solute carrier family 25, member 42 | |
Rabbit | |
35 kDa | |
20 μL | |
Primary | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title