Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A44 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$145.80 - $373.00

Specifications

Antigen SLC25A44
Concentration 0.5 mg/mL
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ABS2238100
View Documents
MilliporeSigma
ABS2238100UL
100 μL
Each for $373.00
Add to cart
 
ABS223825UL
View Documents
MilliporeSigma
ABS223825UL
25 μL
Each for $145.80
Add to cart
 
Description

Description

SLC25A44 Polyclonal antibody specifically detects SLC25A44 in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

SLC25A44
Western Blot
Purified
RUO
Purified rabbit polyclonal antibody in PBS, pH 7.4, containing 0.02% sodium azide.
IgG
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
0.5 mg/mL
Unconjugated
Rabbit
Mouse
KLH-conjugated linear peptides corresponding to 20 amino acids from the N-terminal region, 14 amino acids from the internal region, and 14 amino acids from the C-terminal region of mouse Solute carrier family 25 member 44 (SLC25A44).
Polyclonal
Affinity Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit