Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC25A46 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100184
Description
Antibody detects endogenous levels of total SLC25A46.
SLC25A46 belongs to the SLC25 family of mitochondrial carrier proteins.
Specifications
SLC25A46 | |
Polyclonal | |
Unconjugated | |
SLC25A46 | |
1200007B05Rik; AI325987; HMSN6B; RGD1305072; Slc25a46; Solute carrier family 25 member 46; solute carrier family 25, member 46; TB1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
291709, 67453, 91137 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5EB62, Q96AG3, Q9CQS4 | |
SLC25A46 | |
A synthesized peptide derived from human SLC25A46(Accession Q96AG3), corresponding to amino acid residues G26-P76. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction