Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A46 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
$152.22 - $436.00
Specifications
Antigen | SLC25A46 |
---|---|
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15956520
|
Novus Biologicals
NBP15956520UL |
20 μL |
Each for $152.22
|
|
NBP159565
|
Novus Biologicals
NBP159565 |
100 μL |
Each for $436.00
|
|
Description
SLC25A46 Polyclonal specifically detects SLC25A46 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC25A46 | |
Polyclonal | |
Purified | |
RUO | |
Q96AG3 | |
91137 | |
Synthetic peptides corresponding to SLC25A46(solute carrier family 25, member 46) The peptide sequence was selected from the N terminal of SLC25A46. Peptide sequence RSFSTGSDLGHWVTTPPDIPGSRNLHWGEKSPPYGVPTTSTPYEGPTEEP. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
solute carrier family 25 member 46, solute carrier family 25, member 46, TB1 | |
SLC25A46 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title