Learn More
SLC25A6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148411AP
Catalog No. 50-172-9769
Add to cart
Description
Description
This gene is a member of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this gene functions as a gated pore that translocates ADP from the mitochondrial matrix into the cytoplasm. The protein is implicated in the function of the permability transition pore complex (PTPC), which regulates the release of mitochondrial products that induce apoptosis. The human genome contains several non-transcribed pseudogenes of this gene.
Specifications
Specifications
SLC25A6
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P12236, Q5U680
AAC3, ADP,ATP carrier protein 3, ADP/ATP translocase 3, ANT 2, ANT 3, ANT3, ANT3Y, SLC25A6
Rabbit
IgG
150 μL
-20°C
Primary
293, 67582
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
SLC25A6
Liquid
Slc25a26, SLC25A6
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse