Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC26A1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15940820UL
Description
SLC26A1 Polyclonal specifically detects SLC26A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC26A1 | |
Polyclonal | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Q9H2B4 | |
SLC26A1 | |
Synthetic peptides corresponding to SLC26A1(solute carrier family 26 (sulfate transporter), member 1) The peptide sequence was selected from the C terminal of SLC26A1. Peptide sequence LYSLTGLDAGCMAARRKEGGSETGVGEGGPAQGEDLGPVSTRAALVPAAA. | |
20 μL | |
Primary | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
EDM4, SAT1, SAT-1sulfate anion transporter 1, solute carrier family 26 (sulfate transporter), member 1, Solute carrier family 26 member 1, sulfate anion tranporter AT1, sulfate transporter, sulfate/anion transporter SAT-1 protein | |
Rabbit | |
Affinity Purified | |
RUO | |
10861 | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title