Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC26A1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen SLC26A1
Immunogen Synthetic peptides corresponding to SLC26A1 (solute carrier family 26 (sulfate transporter), member 1) The peptide sequence was selected from the middle region of SLC26A1)(50ug). Peptide sequence ACCSPPVRDILSRGGFLGEGPGDTAEEEQLFLSVHDAVQTARAR
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15965220 View Documents Novus BiologicalsSupplier Diversity Partner
NBP15965220UL
20ul Each for $149.00
Add to cart
 
NBP159652 View Documents Novus BiologicalsSupplier Diversity Partner
NBP159652
100 ul Each for $329.00
Add to cart
 
Description

Description

SLC26A1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blotting.
Specifications

Specifications

SLC26A1
Rabbit
Polyclonal
Unconjugated
10861
Affinity Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
RUO
Synthetic peptides corresponding to SLC26A1 (solute carrier family 26 (sulfate transporter), member 1) The peptide sequence was selected from the middle region of SLC26A1)(50ug). Peptide sequence ACCSPPVRDILSRGGFLGEGPGDTAEEEQLFLSVHDAVQTARAR
Primary
IgG
EDM4, SAT1, SAT-1sulfate anion transporter 1, solute carrier family 26 (sulfate transporter), member 1, Solute carrier family 26 member 1, sulfate anion tranporter AT1, sulfate transporter, sulfate/anion transporter SAT-1 protein
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit