Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC26A5 Polyclonal Antibody
SLC26A5 Polyclonal Antibody
Supplier: Thermo Scientific PA5103158
Description
SLC26A5 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
SLC26A5 | |
Polyclonal | |
Unconjugated | |
SLC26A5 | |
DFNB61; MGC118886; MGC118887; MGC118888; MGC118889; outer hair cell motor protein; PRES; Prestin; prestin (motor protein); Slc26a5; solute carrier family 26 (anion exchanger), member 5; solute carrier family 26 member 5; solute carrier family 26, member 5; solute carrier family 26, member 5 (prestin); solute carrier family 26, member 5 (prestin) isoform 1; solute carrier family 26, member 5 (prestin) isoform 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
375611, 80979, 83819 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P58743, Q99NH7, Q9EPH0 | |
SLC26A5 | |
A synthesized peptide derived from human SLC26A5(Accession P58743), corresponding to amino acid residues A427-V477. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction