Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC28A3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP288294

Catalog No. NB035916


Add to cart

Description

Description

SLC28A3 Polyclonal specifically detects SLC28A3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SLC28A3
Unconjugated
PBS, 2% Sucrose
CNT 3, CNT3concentrative Na+-nucleoside cotransporter, Concentrative Na(+)-nucleoside cotransporter 3, hCNT3, solute carrier family 28 (sodium-coupled nucleoside transporter), member 3, solute carrier family 28 member 3
The immunogen is a synthetic peptide directed towards the middle region of human SLC28A3
100 μg
Cancer
Primary
64078
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Immunogen affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit