Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC29A3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Specifications

Antigen SLC29A3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB028808
View Documents
Novus Biologicals
NBP293466100UL
100 μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
NB028809
View Documents
Novus Biologicals
NBP29346620UL
20 μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

SLC29A3 Polyclonal antibody specifically detects SLC29A3 in Human samples. It is validated for Western Blot
Specifications

Specifications

SLC29A3
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
55315
IgG
Affinity purified
Western Blot
Unconjugated
Rabbit
Human
equilibrative nucleoside transporter 3, ENT3, HCLAP, HJCD, PHID, solute carrier family 29 (nucleoside transporters), member 3
Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human SLC29A3 (NP_060814.4).
Primary
Store at -20°:C. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit