Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC2A12 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550504

Catalog No. PA550504


Add to cart

Description

Description

The antibody detects endogenous levels of total SLC2A12 protein.

Solute carrier family 2, facilitated glucose transporter member 12 is a protein that in humans is encoded by the SLC2A12 gene. SLC2A12 belongs to a family of transporters that catalyze the uptake of sugars through facilitated diffusion. This family of transporters show conservation of 12 transmembrane helices as well as functionally significant amino acid residues.
Specifications

Specifications

SLC2A12
Polyclonal
Unconjugated
SLC2A12
Liquid
SLC2A12
Synthetic peptide corresponding to residues near the C terminal of human solute carrier family 2 (facilitated glucose transporter), member 12
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.7 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q8TD20
GLUT12, GLUT8
Rabbit
IgG
100 μL
-20°C
Primary
154091
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit