Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC2A13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140232
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PESPRWLIQK GQTQKARRIL SQMRGNQTID EEYDSIKNNI EEEEKEVGSA GPVICRMLSY P.
SLC2A13 (solute carrier family 2 (facilitated glucose transporter), member 13) is a 648 multi-pass membrane protein that belongs to the major facilitator superfamily and the sugar transporter family. SLC2A13 is a H+-myo-inositol cotransporter that can also transport related stereoisomers. Human SLC2A13 shares 90% identity with its rat homolog. The rat sequence has 12 transmembrane domains, 3 N-glycosylation sites, several motifs important for glucose transport activity, and an endoplasmic reticulum retention signal and dileucine internalization signal in the N-terminal region. SLC2A13 has been shown to specifically transport myoinositol but not hexoses, and decreasing the pH of the extracellular medium from 7.0 to 5.0 markedly increased transport activity.
Specifications
SLC2A13 | |
Polyclonal | |
Unconjugated | |
SLC2A13 | |
6530403A04; A630029G22Rik; AI505012; Gm308; h(+)-myo-inositol cotransporter; H(+)-myo-inositol symporter; Hmit; proton (H+) myo-inositol symporter; proton myo-inositol cotransporter; proton myo-inositol symporter; SLC2A13; solute carrier family 2 (facilitated glucose transporter), member 13; solute carrier family 2 member 13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114134 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96QE2 | |
SLC2A13 | |
Recombinant protein corresponding to Human SLC2A13. Recombinant protein control fragment (Product #RP-109004). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction