Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC2A13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548621

Catalog No. PA548621

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

H(+)-myo-inositol cotransporter. Can also transport related stereoisomers.
Specifications

Specifications

SLC2A13
Polyclonal
PBS with 0.09% sodium azide
Q96QE2
Proton myo-inositol cotransporter, H(+)-myo-inositol cotransporter, Hmit, H(+)-myo-inositol symporter, Solute carrier family 2 member 13, SLC2A13
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
114134
Western Blot
Unconjugated
SLC2A13
Liquid
SLC2A13
KLH conjugated synthetic peptide between 268-297 amino acids from the Central region of human SLC2A13.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit