Learn More
Aviva Systems Biology Corp SLC30A8 Antibody - middle region (ARP48086_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48086_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC30A8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC30A8. Alternate names: ZnT-8, ZNT8ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59715