Learn More
SLC30A9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA124437
Description
PA1-24437 detects Embryonic Lung Protein in Human samples. PA1-24437 has been successfully used in Western Blot procedures. PA1-24437 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human SLC30A9. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
The gene corresponding to embryonic lung protein [also known as Solute carrier family 30 (Zinc transporter), member 9, SLC30A9] is likely to be an evolutionarily conserved, housekeeping gene that plays a role intimately linked with cellular replication, DNA synthesis and/or transcriptional regulation.
Specifications
| SLC30A9 | |
| Polyclonal | |
| Unconjugated | |
| Q6PML9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10463 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| SLC30A9 | |
| SLC30A9 | |
| Synthetic peptide region of human SLC30A9. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.