Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC34A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568547
Description
This target displays homology in the following species: Dog: 85%; Human: 100%; Mouse: 100%; Rat: 100%.
This gene encodes a member of SLC34A transporter family of proteins, and is expressed primarily in the kidney. It is involved in transporting phosphate into cells via sodium cotransport in the renal brush border membrane, and contributes to the maintenance of inorganic phosphate concentration in the kidney. Mutations in this gene are associated with hereditary hypophosphatemic rickets with hypercalciuria. Alternatively spliced transcript variants varying in the 5' UTR have been found for this gene.
Specifications
SLC34A3 | |
Polyclonal | |
Unconjugated | |
SLC34A3 | |
AI649385; HHRH; Na(+)/Pi cotransporter 2C; Na(+)-dependent phosphate cotransporter 2C; Na+/Pi-cotransporter type IIc; naPi-2c; Npt2c; Npt2c-v1; NptIIc; Slc34a3; Sodium/inorganic phosphate cotransporter IIC; sodium/phosphate cotransporter 2C; Sodium-dependent phosphate transport protein 2C; sodium-dependent phosphate transport protein 2C; LOW QUALITY PROTEIN: sodium-dependent phosphate transport protein 2C; sodium-phosphate cotransporter IIC; sodium-phosphate transport protein 2C; solute carrier family 34 (sodium phosphate), member 3; solute carrier family 34 (type II sodium/phosphate contransporter), member 3; solute carrier family 34 (type II sodium/phosphate cotransporter), member 3; solute carrier family 34 member 3; type IIc Na+/Pi cotransporter; type IIc Na+/Pi-cotransporter; type IIc NaPi-cotransporter-v1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
142680 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N130 | |
SLC34A3 | |
synthetic peptide directed towards the middle region of human SLC34A3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction