Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC35A3 Monoclonal Antibody (4B6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA521475
Description
Peptide Sequence: DSKCSLRALN RVLHDEILNK PMETLKLAIP SGIYTLQNNL LYVALSNLDA ATY.
Uridine diphosphate-N-acetylglucosamine (UDP-GlcNAc) transporter in the Golgi apparatus. May supply UDP-GlcNAc as substrate for Golgi-resident glycosyltransferases that generate branching of diantennary oligosaccharides.
Specifications
SLC35A3 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9Y2D2 | |
SLC35A3 | |
SLC35A3 (NP_036375, 61 a.a. ~ 113 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
4B6 | |
Unconjugated | |
SLC35A3 | |
AMRS; golgi UDP-GlcNAc transporter; Slc35a3; solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member 3; solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member A3; solute carrier family 35 member 3; solute carrier family 35 member 3A; solute carrier family 35 member A3; UDP N-acetylglucosamine transporter; UDP-N-acetylglucosamine transporter | |
Mouse | |
Affinity Chromatography | |
RUO | |
23443 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction