Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC35A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553292
Description
Immunogen sequence: SSRSVLSPVV GTDAPDQHLE LKKPQELKEM ERLPLANEDK TMF Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 37%.
Uridine diphosphate-N-acetylglucosamine (UDP-GlcNAc) transporter in the Golgi apparatus. May supply UDP-GlcNAc as substrate for Golgi-resident glycosyltransferases that generate branching of diantennary oligosaccharides.
Specifications
SLC35A3 | |
Polyclonal | |
Unconjugated | |
SLC35A3 | |
AMRS; golgi UDP-GlcNAc transporter; Slc35a3; solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member 3; solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member A3; solute carrier family 35 member 3; solute carrier family 35 member 3A; solute carrier family 35 member A3; UDP N-acetylglucosamine transporter; UDP-N-acetylglucosamine transporter | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23443 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2D2 | |
SLC35A3 | |
Recombinant protein corresponding to Human SLC35A3. Recombinant protein control fragment (Product #RP-91142). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction