Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC35D3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556473
Description
Immunogen sequence: RKQSNYEDLE AQPRGEEAQL SGDQLPFVME ELPGEGGNGR SEGGEAAGGP AQESRQEVRG SPRGVPLVAG SSEEGSRRSL KDAYLEVWRL VRGTRYM Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 71%.
May play a role in hemostasis as a regulator of the biosynthesis of platelet-dense granules.
Specifications
SLC35D3 | |
Polyclonal | |
Unconjugated | |
SLC35D3 | |
6230421J19Rik; frc, fringe-like 1; Frcl1; Fringe connection-like protein 1; SLC35D3; Solute carrier family 35 member D3; solute carrier family 35, member D3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
340146 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5M8T2 | |
SLC35D3 | |
Recombinant protein corresponding to Human SLC35D3. Recombinant protein control fragment (Product #RP-95017). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction