Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC35E2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551318

Catalog No. PA551318


Add to cart

Description

Description

The antibody detects endogenous levels of total SLC35E2 protein.

Putative transporter.
Specifications

Specifications

SLC35E2
Polyclonal
Unconjugated
SLC35E2
Liquid
SLC35E2
Fusion protein of human SLC35E2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P0CK97
solute carrier family 35 member E2, solute carrier family 35, member E2
Rabbit
IgG
100 μL
-20°C
Primary
9906
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit