Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC36A4 Antibody (PAT-4/9/H10), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250237V
Description
SLC36A4 Monoclonal specifically detects SLC36A4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Knockdown Validated.Specifications
SLC36A4 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SLC36A4 | |
Antigenic amino acid sequence within the N-terminus of SLC36A4 (REELDMDVMRPLINE-C). | |
0.1 mL | |
Cellular Signaling, metabolism | |
120103 | |
Store at 4C in the dark. | |
IgG2a κ |
Western Blot, Immunohistochemistry, KnockDown | |
PAT-4/9/H10 | |
Western Blot, Immunohistochemistry | |
proton/amino acid transporter 4, solute carrier family 36 (proton/amino acid symporter), member 4 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction