Learn More
SLC38A1 Mouse anti-Human, Mouse, Rat, DyLight 650, Clone: S104-32, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259311C
Catalog No. NBP259311C
Add to cart
Description
Description
SLC38A1 Monoclonal antibody specifically detects SLC38A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
SLC38A1
S104-32
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
SLC38A1
Fusion protein amino acids 1- 63 of rat SNAT1
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 50kDa.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Amino acid transporter A1, ATA1SNAT1, NAT2N-system amino acid transporter 2, SAT1amino acid transporter system A1, sodium-coupled neutral amino acid transporter 1, Solute carrier family 38 member 1, solute carrier family 38, member 1, System A amino acid transporter 1, System N amino acid transporter 1
Mouse
IgG1
100 ul
Primary
81539
Human, Mouse, Rat