Learn More
SLC38A1 Mouse anti-Human, Mouse, Rat, FITC, Clone: S104-32, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259311F
Catalog No. NBP259311F
Add to cart
Description
Description
SLC38A1 Monoclonal antibody specifically detects SLC38A1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
SLC38A1
S104-32
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
SLC38A1
Fusion protein amino acids 1- 63 of rat SNAT1
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 50kDa.
Immunohistochemistry, Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
Amino acid transporter A1, ATA1SNAT1, NAT2N-system amino acid transporter 2, SAT1amino acid transporter system A1, sodium-coupled neutral amino acid transporter 1, Solute carrier family 38 member 1, solute carrier family 38, member 1, System A amino acid transporter 1, System N amino acid transporter 1
Mouse
IgG1
100 ul
Primary
81539
Human, Mouse, Rat