Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC38A1 Mouse anti-Human, Rat, Clone: S104-32, Abnova™

Mouse monoclonal antibody raised against human SLC38A1.

Manufacturer:  Abnova Corporation MAB12695

Catalog No. 89-934-936


Add to cart

Description

Description

Amino acid transporters play essential roles in the uptake of nutrients, production of energy, chemical metabolism, detoxification, and neurotransmitter cycling. SLC38A1 is an important transporter of glutamine, an intermediate in the detoxification of ammonia and the production of urea. Glutamine serves as a precursor for the synaptic transmitter, glutamate (Gu et al., 2001 [PubMed 11325958]).[supplied by OMIM]

Specifications

Specifications

SLC38A1
S104-32
Mouse monoclonal antibody raised against human SLC38A1.
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
ATA1/NAT2/SAT1/SNAT1
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Rat
Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunocytochemistry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
SLC38A1
Recombinant fusion protein corresponding to amino acids 1-63 of human SLC38A1.
Protein G purification
RUO
Primary
81539
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit