Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC38A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117341
Description
Centrifuge briefly prior to opening.
Amino acid transporters play essential roles in the uptake of nutrients, production of energy, chemical metabolism, detoxification, and neurotransmitter cycling. SLC38A1 is an important transporter of glutamine, an intermediate in the detoxification of ammonia and the production of urea. Glutamine serves as a precursor for the synaptic transmitter, glutamate.
Specifications
SLC38A1 | |
Polyclonal | |
Unconjugated | |
SLC38A1 | |
AA408026; AA409865; AL022800; Amino acid transporter A1; amino acid transporter system A1; ATA1; AU015942; GlnT; glutamine transporter; MNat2; Nat2; N-system amino acid transporter 2; rATA1; S38A1; Sa2; SAT1; SLC38A1; SNAT 1; Snat1; sodium-coupled neutral amino acid transporter 1; Solute carrier family 38 member 1; solute carrier family 38, member 1; system A amino acid transporter 1; System A transporter 2; system N amino acid transporter 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
81539 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q9H2H9 | |
SLC38A1 | |
Recombinant protein encompassing a sequence within the N-terminus region of human SLC38A1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction