Learn More
SLC39A5 Mouse anti-Human, Clone: OTI2F2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810144
Catalog No. 50-166-9912
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The zinc transporter ZIP5, also known as SLC39A5, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP5 is closely related to ZIP4, and is found in many of the same tissues, including intestine, pancreas, liver, and kidney, but unlike ZIP4, ZIP5 localizes to the basolateral membrane in kidney cells and its activity is not downregulated by zinc repletion. Recent evidence suggests that ZIP4 and ZIP5 are reciprocally regulated, with translation of ZIP5 mRNA decreasing and ZIP5 protein being rapidly internalized and degraded in response to zinc deficiency.
Specifications
Specifications
SLC39A5
Monoclonal
Unconjugated
SLC39A5
Lyophilized
SLC39A5
Human recombit protein fragment corresponding to amino acids 48-178 of human SLC39A5 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI2F2
PBS with 8% trehalose and no preservative; pH 7.3
Q6ZMH5
LZT-Hs7, MYP24, ZIP5
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
283375