Learn More
SLC3A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 163431AP
Catalog No. 50-173-0883
Add to cart
Description
Description
This gene encodes a type II membrane glycoprotein which is one of the components of the renal amino acid transporter which transports neutral and basic amino acids in the renal tubule and intestinal tract. Mutations and deletions in this gene are associated with cystinuria. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
Specifications
SLC3A1
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q07837, Q64319, Q91WV7
ATR1, CSNU1, D2H, NBAT, RBAT, SLC3A1
Rabbit
IgG
150 μL
-20°C
Primary
20532, 29484, 6519
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
SLC3A1
Liquid
SLC3A1
SLC3A1 Fusion Protein Ag9525
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat